Assay Details
| Gene Symbol: CA3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277410.1 | NP_001264339.1 | 5 - 6 | 655 | 131 | |
| GenBank mRNA | CR352666.1 | - | 5 - 6 | 594 | 131 | |
More Information
Set Membership: |
|

CA3B
Genomic Map