Assay Details
| Gene Symbol: ZNF280D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943779.3 | XP_004943836.3 | 10 - 11 | 1663 | 77 | |
| XM_004943784.3 | XP_004943841.3 | 10 - 11 | 1664 | 77 | ||
| XM_015291869.2 | XP_015147355.2 | 8 - 9 | 1288 | 77 | ||
| XM_413961.6 | XP_413961.6 | 9 - 10 | 1595 | 77 | ||
| GenBank mRNA | CR352765.1 | - | 4 - 5 | 588 | 77 | |
More Information
Set Membership: |
|

ZNF280D
Genomic Map