Assay Details
| Gene Symbol: COX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX932336.1 | - | 3 - 4 | 362 | 97 | |
| CR353996.1 | - | 3 - 4 | 364 | 97 | ||
| CR405829.1 | - | 3 - 4 | 343 | 97 | ||
More Information
Set Membership: |
|

COX16
Genomic Map