Assay Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302125.1 | NP_001289054.1 | 1 - 2 | 290 | 66 | |
| GenBank mRNA | CR354038.1 | - | 1 - 2 | 290 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302125.1 transcripts.
Set Membership: |
|

LSM6
Genomic Map