Assay Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302124.1 | NP_001289053.1 | 1 - 2 | 471 | 143 | |
| NM_001302125.1 | NP_001289054.1 | 2 - 3 | 402 | 143 | ||
| NM_001302126.1 | NP_001289055.1 | 2 - 3 | 130 | 143 | ||
| GenBank mRNA | BU279150.1 | - | 1 - 2 | 471 | 143 | |
| BU390777.1 | - | 2 - 3 | 130 | 143 | ||
| CR353169.1 | - | 1 - 2 | 129 | 143 | ||
| CR354038.1 | - | 2 - 3 | 402 | 143 | ||
| CR390394.1 | - | 1 - 2 | 411 | 143 | ||
More Information
Set Membership: |
|

LSM6
Genomic Map