Assay Details
| Gene Symbol: PPP1R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420586.6 | XP_420586.2 | 1 - 2 | 58 | 76 | |
| GenBank mRNA | BX929364.2 | - | 1 - 2 | 48 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_420586.6 transcripts.
Set Membership: |
|

PPP1R3B
Genomic Map