Assay Details
| Gene Symbol: PRAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276439.1 | XP_015131925.1 | 6 | 139 | ||
| XM_015276440.2 | XP_015131926.1 | 7 | 139 | |||
| XM_015276441.2 | XP_015131927.1 | 6 | 139 | |||
| XM_015276442.2 | XP_015131928.1 | 6 | 139 | |||
| XM_025150156.1 | XP_025005924.1 | 6 | 139 | |||
| GenBank mRNA | BX929590.2 | - | 1 | 1146 | 140 | |
| BX932561.2 | - | 1 | 1151 | 138 | ||
| BX934417.1 | - | 2 | 138 | |||
More Information
Set Membership: |
|

PRAG1
Genomic Map