Assay Details
| Gene Symbol: GK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640510.4 | XP_003640558.2 | 6 - 7 | 677 | 86 | |
| XM_003640511.4 | XP_003640559.2 | 6 - 7 | 677 | 86 | ||
| XM_015302211.2 | XP_015157697.1 | 6 - 7 | 677 | 86 | ||
| XM_015302218.2 | XP_015157704.1 | 6 - 7 | 677 | 86 | ||
| GenBank mRNA | BX929694.2 | - | 6 - 7 | 584 | 86 | |
More Information
Set Membership: |
|

GK2
Genomic Map