Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302168.1 | NP_001289097.1 | 1 - 2 | 601 | 60 | |
| GenBank mRNA | BX929857.2 | - | 1 - 2 | 611 | 60 | |
More Information
Set Membership: |
|

CCL19
Genomic Map