Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293203.1 | NP_001280132.1 | 2 - 3 | 121 | 69 | |
| GenBank mRNA | BU276503.1 | - | 2 - 3 | 121 | 69 | |
| BX930131.2 | - | 2 - 3 | 96 | 69 | ||
| BX950793.2 | - | 2 - 3 | 308 | 69 | ||
More Information
Set Membership: |
|

SORD
Genomic Map