Assay Details
| Gene Symbol: PLPPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290742.2 | XP_015146228.1 | 5 - 6 | 758 | 73 | |
| XM_015290743.2 | XP_015146229.1 | 2 - 3 | 486 | 73 | ||
| XM_015290744.2 | XP_015146230.1 | 3 - 4 | 1189 | 73 | ||
| XM_025152941.1 | XP_025008709.1 | 2 - 3 | 486 | 73 | ||
| XM_422320.5 | XP_422320.2 | 2 - 3 | 486 | 73 | ||
| GenBank mRNA | BX930216.2 | - | 2 - 3 | 410 | 73 | |
More Information
Set Membership: |
|

PLPPR5
Genomic Map