Assay Details
| Gene Symbol: USMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252029.1 | NP_001238958.1 | 2 - 3 | 87 | 76 | |
| GenBank mRNA | BX929561.2 | - | 2 - 3 | 87 | 76 | |
| BX930500.2 | - | 2 - 3 | 95 | 76 | ||
| CR407460.1 | - | 1 - 2 | 610 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252029.1 transcripts.
Set Membership: |
|

USMG5
Genomic Map