Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417013.5 | XP_417013.5 | 2 - 3 | 1281 | 76 | |
| GenBank mRNA | BX930662.2 | - | 1 - 2 | 58 | 76 | |
| BX935940.2 | - | 1 - 2 | 433 | 76 | ||
More Information
Set Membership: |
|

KLF5
Genomic Map