Assay Details
| Gene Symbol: DLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142254.1 | NP_001135726.1 | 1 - 2 | 98 | 59 | |
| XM_025150792.1 | XP_025006560.1 | 5 - 6 | 879 | 59 | ||
| GenBank mRNA | BX930935.2 | - | 1 - 2 | 89 | 59 | |
| EU288039.1 | - | 1 - 2 | 98 | 59 | ||
More Information
Set Membership: |
|

DLK1
Genomic Map