Assay Details
| Gene Symbol: PINX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149179.1 | XP_025004947.1 | 5 - 6 | 405 | 104 | |
| XM_025149180.1 | XP_025004948.1 | 5 - 6 | 406 | 104 | ||
| XM_420036.6 | XP_420036.3 | 5 - 6 | 413 | 104 | ||
| GenBank mRNA | BX932109.2 | - | 5 - 6 | 422 | 104 | |
More Information
Set Membership: |
|

PINX1
Genomic Map