Assay Details
| Gene Symbol: PTTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277740.1 | NP_001264669.1 | 5 - 6 | 756 | 138 | |
| NM_001277741.1 | NP_001264670.1 | 4 - 5 | 726 | 138 | ||
| XM_025154831.1 | XP_025010599.1 | 5 - 6 | 687 | 138 | ||
| GenBank mRNA | AJ441614.1 | - | 5 - 6 | 589 | 138 | |
| BU291226.1 | - | 4 - 5 | 581 | 138 | ||
| BX932212.2 | - | 5 - 6 | 655 | 138 | ||
More Information
Set Membership: |
|

PTTG2
Genomic Map