Assay Details
| Gene Symbol: TCF21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277711.1 | NP_001264640.1 | 1 - 2 | 617 | 68 | |
| GenBank mRNA | BX932800.2 | - | 1 - 2 | 618 | 68 | |
More Information
Set Membership: |
|

TCF21
Genomic Map