Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277489.1 | NP_001264418.1 | 1 - 2 | 92 | ||
| GenBank mRNA | BU290618.1 | - | 1 - 2 | 92 | ||
| BX933405.2 | - | 1 - 2 | 92 | |||
| BX934100.2 | - | 1 - 2 | 81 | 92 | ||
More Information
Set Membership: |
|

NIP7
Genomic Map