Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941425.3 | XP_004941482.1 | 1 - 2 | 1174 | 72 | |
XM_004941426.3 | XP_004941483.1 | 1 - 2 | 1066 | 72 | ||
XM_004941428.3 | XP_004941485.1 | 1 - 2 | 1174 | 72 | ||
XM_004941431.3 | XP_004941488.1 | 1 - 2 | 1174 | 72 | ||
XM_015286658.2 | XP_015142144.1 | 1 - 2 | 1174 | 72 | ||
GenBank mRNA | BX933945.2 | - | 1 - 2 | 586 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
