Assay Details
| Gene Symbol: GALNT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199584.1 | NP_001186513.1 | 2 - 3 | 497 | 64 | |
| XM_015286372.2 | XP_015141858.1 | 3 - 4 | 681 | 64 | ||
| XM_015286373.2 | XP_015141859.1 | 6 - 7 | 2487 | 64 | ||
| XM_025150668.1 | XP_025006436.1 | 3 - 4 | 686 | 64 | ||
| XM_025150669.1 | XP_025006437.1 | 6 - 7 | 2487 | 64 | ||
| GenBank mRNA | BX934209.2 | - | 2 - 3 | 497 | 64 | |
More Information
Set Membership: |
|

GALNT18
Genomic Map