Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424710.5 | XP_424710.2 | 3 - 4 | 670 | 104 | |
| GenBank mRNA | BX929746.1 | - | 2 - 3 | 178 | 104 | |
| BX933208.1 | - | 3 - 4 | 386 | 104 | ||
| BX934335.2 | - | 1 - 2 | 34 | 104 | ||
| BX935761.1 | - | 3 - 4 | 307 | 104 | ||
| CR386784.1 | - | 2 - 3 | 296 | 104 | ||
More Information
Set Membership: |
|

RFESD
Genomic Map