Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231830.5 | XP_001231831.3 | 2 - 3 | 442 | 73 | |
| XM_004939811.3 | XP_004939868.1 | 2 - 3 | 536 | 73 | ||
| XM_004939812.3 | XP_004939869.1 | 2 - 3 | 492 | 73 | ||
| XM_015277995.2 | XP_015133481.2 | 2 - 3 | 1863 | 73 | ||
| XM_015277996.2 | XP_015133482.1 | 2 - 3 | 357 | 73 | ||
| XM_015277997.2 | XP_015133483.1 | 2 - 3 | 209 | 73 | ||
| XM_015277999.2 | XP_015133485.1 | 2 - 3 | 209 | 73 | ||
| XM_015278000.2 | XP_015133486.1 | 2 - 3 | 290 | 73 | ||
| XM_025147908.1 | XP_025003676.1 | 2 - 3 | 208 | 73 | ||
| GenBank mRNA | BX934672.2 | - | 2 - 3 | 447 | 73 | |
More Information
Set Membership: |
|

KCTD1
Genomic Map