Assay Details
| Gene Symbol: TCP11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199407.1 | NP_001186336.1 | 3 - 4 | 326 | 86 | |
| XM_015286317.2 | XP_015141803.1 | 2 - 3 | 261 | 86 | ||
| XM_025150633.1 | XP_025006401.1 | 3 - 4 | 499 | 86 | ||
| GenBank mRNA | BX935700.2 | - | 3 - 4 | 326 | 86 | |
More Information
Set Membership: |
|

TCP11L1
Genomic Map