Assay Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147970.1 | XP_025003738.1 | 1 - 2 | 509 | 77 | |
| XM_419214.6 | XP_419214.5 | 1 - 2 | 502 | 77 | ||
| GenBank mRNA | BX936021.2 | - | 1 - 2 | 473 | 77 | |
More Information
Set Membership: |
|

IMPAD1
Genomic Map