Assay Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277757.1 | NP_001264686.1 | 6 - 7 | 577 | 75 | |
| NM_001277758.1 | NP_001264687.1 | 4 - 5 | 466 | 75 | ||
| XM_025150040.1 | XP_025005808.1 | 6 - 7 | 464 | 75 | ||
| GenBank mRNA | BU323063.1 | - | 6 - 7 | 577 | 75 | |
| BU377256.1 | - | 2 - 3 | 169 | 75 | ||
| CR353977.1 | - | 6 - 7 | 509 | 75 | ||
| CR354137.1 | - | 4 - 5 | 422 | 75 | ||
More Information
Set Membership: |
|

MRPS9
Genomic Map