Assay Details
| Gene Symbol: TMEM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284918.2 | XP_015140404.1 | 4 - 5 | 318 | 109 | |
| XM_015284919.2 | XP_015140405.1 | 3 - 4 | 437 | 109 | ||
| XM_419915.6 | XP_419915.3 | 4 - 5 | 316 | 109 | ||
| GenBank mRNA | BX929565.1 | - | 6 - 7 | 583 | 109 | |
| BX930672.2 | - | 4 - 5 | 298 | 109 | ||
| CR385154.1 | - | 5 - 6 | 413 | 109 | ||
| CR405747.1 | - | 4 - 5 | 272 | 109 | ||
More Information
Set Membership: |
|

TMEM14A
Genomic Map