Assay Details
| Gene Symbol: LSM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418245.6 | XP_418245.3 | 4 - 5 | 372 | 78 | |
| GenBank mRNA | CR386221.1 | - | 1 - 2 | 35 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSM4| Gene Symbol: LSM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418245.6 | XP_418245.3 | 4 - 5 | 372 | 78 | |
| GenBank mRNA | CR386221.1 | - | 1 - 2 | 35 | 78 | |
Set Membership: |
|