Assay Details
Gene Symbol: AMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004940779.3 | XP_004940836.1 | 2 - 3 | 4283 | 151 | |
XM_004940780.3 | XP_004940837.1 | 2 - 3 | 4579 | 151 | ||
XM_015278574.2 | XP_015134060.1 | 2 - 3 | 3642 | 151 | ||
GenBank mRNA | CR386322.1 | - | 2 - 3 | 1244 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
