Assay Details
| Gene Symbol: GUCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197038.1 | NP_001183967.1 | 3 | 1108 | 112 | |
| GenBank mRNA | CR386770.1 | - | 1 | 649 | 112 | |
More Information
Set Membership: |
|

GUCA2B
Genomic Map