Assay Details
| Gene Symbol: SIAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292416.2 | XP_015147902.1 | 3 | 1076 | 91 | |
| XM_015292417.2 | XP_015147903.1 | 3 | 879 | 91 | ||
| XM_015292418.2 | XP_015147904.1 | 2 | 766 | 91 | ||
| XM_015292419.2 | XP_015147905.1 | 2 | 774 | 91 | ||
| GenBank mRNA | CR386772.1 | - | 1 | 647 | 91 | |
More Information
Set Membership: |
|

SIAH1
Genomic Map