Assay Details
| Gene Symbol: DTX3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289955.2 | XP_015145441.1 | 2 - 3 | 417 | 65 | |
| XM_422114.6 | XP_422114.4 | 2 - 3 | 394 | 65 | ||
| GenBank mRNA | CR387363.1 | - | 1 - 2 | 97 | 65 | |
More Information
Set Membership: |
|

DTX3L
Genomic Map