Assay Details
| Gene Symbol: WRNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418979.6 | XP_418979.2 | 4 - 5 | 1260 | 62 | |
| GenBank mRNA | AB104724.1 | - | 3 - 4 | 555 | 62 | |
More Information
Set Membership: |
|

WRNIP1
Genomic Map