Assay Details
| Gene Symbol: SCN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293281.1 | NP_001280210.1 | 6 - 7 | 977 | 88 | |
| XM_004942786.3 | XP_004942843.1 | 7 - 8 | 1076 | 88 | ||
| XM_004942787.2 | XP_004942844.1 | 7 - 8 | 1309 | 88 | ||
| XM_015289631.1 | XP_015145117.1 | 7 - 8 | 1309 | 88 | ||
| XM_015289632.2 | XP_015145118.1 | 7 - 8 | 1309 | 88 | ||
| XM_015289633.1 | XP_015145119.1 | 7 - 8 | 1309 | 88 | ||
| XM_015289634.1 | XP_015145120.1 | 7 - 8 | 1310 | 88 | ||
| XM_015289635.2 | XP_015145121.1 | 7 - 8 | 1310 | 88 | ||
| GenBank mRNA | AJ001490.1 | - | 3 - 4 | 390 | 88 | |
More Information
Set Membership: |
|

SCN2A
Genomic Map