Assay Details
| Gene Symbol: SHQ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944692.2 | XP_004944749.1 | 6 - 7 | 862 | 92 | |
| XM_004944693.3 | XP_004944750.1 | 6 - 7 | 671 | 92 | ||
| XM_015293230.1 | XP_015148716.1 | 6 - 7 | 877 | 92 | ||
| XM_015293231.1 | XP_015148717.1 | 6 - 7 | 862 | 92 | ||
| XM_025154767.1 | XP_025010535.1 | 6 - 7 | 737 | 92 | ||
| XM_025154768.1 | XP_025010536.1 | 6 - 7 | 862 | 92 | ||
| XM_414429.6 | XP_414429.4 | 6 - 7 | 732 | 92 | ||
| GenBank mRNA | AJ719613.1 | - | 6 - 7 | 732 | 92 | |
More Information
Set Membership: |
|

SHQ1
Genomic Map