Assay Details
| Gene Symbol: ABI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039281.2 | NP_001034370.2 | 1 - 2 | 241 | 67 | |
| GenBank mRNA | AJ720766.1 | - | 1 - 2 | 194 | 67 | |
More Information
Set Membership: |
|

ABI1
Genomic Map