Assay Details
| Gene Symbol: PDIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419952.6 | XP_419952.1 | 2 - 3 | 231 | 78 | |
| GenBank mRNA | AJ851428.1 | - | 2 - 3 | 246 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDIA6| Gene Symbol: PDIA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419952.6 | XP_419952.1 | 2 - 3 | 231 | 78 | |
| GenBank mRNA | AJ851428.1 | - | 2 - 3 | 246 | 78 | |
Set Membership: |
|