Assay Details
| Gene Symbol: MMP23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947398.3 | XP_004947455.1 | 2 - 3 | 2217 | 68 | |
| XM_025142634.1 | XP_024998402.1 | 2 - 3 | 235 | 68 | ||
| XM_417569.4 | XP_417569.1 | 2 - 3 | 718 | 68 | ||
| GenBank mRNA | BX929570.1 | - | 2 - 3 | 162 | 68 | |
| BX934969.1 | - | 2 - 3 | 307 | 68 | ||
More Information
Set Membership: |
|

MMP23A
Genomic Map