Assay Details
| Gene Symbol: AADAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943567.3 | XP_004943624.1 | 3 - 4 | 2094 | 126 | |
| XM_015291844.1 | XP_015147330.1 | 4 - 5 | 2249 | 126 | ||
| XM_025153460.1 | XP_025009228.1 | 3 - 4 | 2094 | 126 | ||
| XM_025153461.1 | XP_025009229.1 | 3 - 4 | 2090 | 126 | ||
| XM_422836.6 | XP_422836.2 | 3 - 4 | 485 | 126 | ||
| GenBank mRNA | BX929777.1 | - | 3 - 4 | 441 | 126 | |
| BX931204.1 | - | 3 - 4 | 475 | 126 | ||
More Information
Set Membership: |
|

AADAC
Genomic Map