Assay Details
| Gene Symbol: AGPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419916.6 | XP_419916.1 | 4 - 5 | 597 | 67 | |
| GenBank mRNA | BX930094.1 | - | 4 - 5 | 402 | 67 | |
| BX930171.1 | - | 4 - 5 | 523 | 67 | ||
More Information
Set Membership: |
|

AGPAT5
Genomic Map