Assay Details
| Gene Symbol: TROVE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943131.3 | XP_004943188.1 | 6 - 7 | 1263 | 75 | |
| XM_422201.4 | XP_422201.1 | 6 - 7 | 1413 | 75 | ||
| GenBank mRNA | BX931878.1 | - | 3 - 4 | 333 | 75 | |
| BX950771.2 | - | 5 - 6 | 1199 | 75 | ||
More Information
Set Membership: |
|

TROVE2
Genomic Map