Assay Details
| Gene Symbol: RAB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935486.3 | XP_004935543.1 | 9 | 1842 | 75 | |
| XM_419573.6 | XP_419573.1 | 8 | 1834 | 75 | ||
| GenBank mRNA | CR388569.1 | - | 1 | 750 | 75 | |
More Information
Set Membership: |
|

RAB4A
Genomic Map