Assay Details
| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_413901.6 | XP_413901.3 | 7 - 8 | 942 | 92 | |
| GenBank mRNA | CR388708.1 | - | 6 - 7 | 681 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTFMT| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_413901.6 | XP_413901.3 | 7 - 8 | 942 | 92 | |
| GenBank mRNA | CR388708.1 | - | 6 - 7 | 681 | 92 | |
Set Membership: |
|