Assay Details
| Gene Symbol: TOMM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_430449.5 | XP_430449.2 | 1 - 2 | 198 | 109 | |
| GenBank mRNA | CR388998.1 | - | 1 - 2 | 186 | 109 | |
More Information
Set Membership: |
|

TOMM5
Genomic Map