Assay Details
| Gene Symbol: CENPV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415846.6 | XP_415846.2 | 1 - 2 | 392 | 72 | |
| GenBank mRNA | CR352613.1 | - | 1 - 2 | 230 | 72 | |
| CR389739.1 | - | 1 - 2 | 240 | 72 | ||
More Information
Set Membership: |
|

CENPV
Genomic Map