Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422291.4 | XP_422291.1 | 2 - 3 | 270 | 72 | |
| GenBank mRNA | CR353953.1 | - | 2 - 3 | 228 | 72 | |
| CR390707.1 | - | 2 - 3 | 204 | 72 | ||
| CR523147.1 | - | 2 - 3 | 244 | 72 | ||
More Information
Set Membership: |
|

TSEN15
Genomic Map