Assay Details
| Gene Symbol: EXOC3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421382.6 | XP_421382.4 | 10 - 11 | 2567 | 100 | |
| GenBank mRNA | CR390736.1 | - | 2 - 3 | 194 | 100 | |
More Information
Set Membership: |
|

EXOC3L4
Genomic Map