Assay Details
| Gene Symbol: PAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278707.2 | XP_015134193.1 | 10 - 11 | 1845 | 76 | |
| XM_015278710.2 | XP_015134196.1 | 10 - 11 | 1844 | 76 | ||
| XM_015278716.2 | XP_015134202.1 | 10 - 11 | 1845 | 76 | ||
| XM_015278721.2 | XP_015134207.1 | 9 - 10 | 1783 | 76 | ||
| XM_015278725.2 | XP_015134211.1 | 9 - 10 | 1782 | 76 | ||
| XM_015278729.2 | XP_015134215.1 | 9 - 10 | 1703 | 76 | ||
| XM_015278731.2 | XP_015134217.1 | 9 - 10 | 1684 | 76 | ||
| XM_015278733.2 | XP_015134219.1 | 8 - 9 | 1640 | 76 | ||
| XM_015278741.2 | XP_015134227.1 | 8 - 9 | 1621 | 76 | ||
| XM_015278744.2 | XP_015134230.1 | 8 - 9 | 1620 | 76 | ||
| XM_015278750.2 | XP_015134236.1 | 10 - 11 | 1846 | 76 | ||
| XM_015278755.2 | XP_015134241.1 | 7 - 8 | 1478 | 76 | ||
| XM_015278762.1 | XP_015134248.1 | 8 - 9 | 891 | 76 | ||
| XM_025146837.1 | XP_025002605.1 | 10 - 11 | 1846 | 76 | ||
| XM_025146838.1 | XP_025002606.1 | 10 - 11 | 1846 | 76 | ||
| GenBank mRNA | CR391044.1 | - | 2 - 3 | 316 | 76 | |
More Information
Set Membership: |
|

PAN3
Genomic Map