Assay Details
| Gene Symbol: PPP2R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936921.3 | XP_004936978.1 | 4 - 5 | 3079 | 68 | |
| XM_004936922.3 | XP_004936979.1 | 4 - 5 | 3078 | 68 | ||
| XM_004936923.3 | XP_004936980.1 | 3 - 4 | 696 | 68 | ||
| XM_015291357.2 | XP_015146843.1 | 4 - 5 | 3094 | 68 | ||
| XM_422556.6 | XP_422556.4 | 4 - 5 | 3076 | 68 | ||
| GenBank mRNA | CR391262.1 | - | 3 - 4 | 415 | 68 | |
More Information
Set Membership: |
|

PPP2R3A
Genomic Map