Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289355.2 | XP_015144841.1 | 4 | 1673 | 68 | |
| GenBank mRNA | CR391695.1 | - | 1 | 747 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ORMDL1| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289355.2 | XP_015144841.1 | 4 | 1673 | 68 | |
| GenBank mRNA | CR391695.1 | - | 1 | 747 | 68 | |
Set Membership: |
|