Assay Details
| Gene Symbol: KIAA1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148358.1 | XP_025004126.1 | 3 | 3933 | 109 | |
| XM_025148359.1 | XP_025004127.1 | 3 | 1585 | 109 | ||
| XM_025148360.1 | XP_025004128.1 | 3 | 1659 | 109 | ||
| XM_418578.6 | XP_418578.5 | 3 | 3933 | 109 | ||
| GenBank mRNA | CR405729.1 | - | 1 | 252 | 109 | |
| CR405765.1 | - | 1 | 271 | 109 | ||
| CR407354.1 | - | 1 | 439 | 109 | ||
More Information
Set Membership: |
|

KIAA1462
Genomic Map